Product Details

SNP ID
rs61757610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106924032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATACTTTGCAAGACAGGCCCTT[C/T]GCACAGTCGCAACGCTGGAAAATTT
Phenotype
MIM: 605415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKK2 PubMed Links

Gene Details

Gene
DKK2
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014421.2 1407 Silent Mutation GCA,GCG A234A NP_055236.1

View Full Product Details