Product Details

SNP ID
rs57552844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139504448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAAATACTTTTGGAGCAGAAA[A/C]CCCTTTTCTGTTTTTCCTTTTTTTT
Phenotype
MIM: 606594
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SETD7 PubMed Links

Gene Details

Gene
SETD7
Gene Name
SET domain containing lysine methyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306199.1 Intron NP_001293128.1
NM_001306200.1 Intron NP_001293129.1
NM_030648.3 Intron NP_085151.1
XM_017008661.1 Intron XP_016864150.1

View Full Product Details