Product Details

SNP ID
hCV89763649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108562728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCGGCCCCGGCCCGACTCTAT[C/T]ACAAGAAGGTAGGGACAAAAGAGGG
Phenotype
MIM: 611911 MIM: 611684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISCU PubMed Links

Gene Details

Gene
ISCU
Gene Name
iron-sulfur cluster assembly enzyme
There are no transcripts associated with this gene.

Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
There are no transcripts associated with this gene.

View Full Product Details