Product Details

SNP ID
hCV89791340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:207496275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATGTGCTTGGGGAGAATGGGGG[C/T]CTCTTCTCCAAGAAGCCCGGAGCCT
Phenotype
MIM: 120620 MIM: 120650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000573.3 148 Missense Mutation GCC,GTC A3V NP_000564.2
NM_000651.4 148 Missense Mutation GCC,GTC A3V NP_000642.3
Gene
CR2
Gene Name
complement component 3d receptor 2
There are no transcripts associated with this gene.

View Full Product Details