Product Details

SNP ID
hCV89800748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:207506789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAGGCATCCAGTTCGGATCCC[A/G]AATTAAATATTCTTGTACTAAAGGG
Phenotype
MIM: 120620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CR1 PubMed Links

Gene Details

Gene
CR1
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000573.3 517 Missense Mutation CAA,CGA Q126R NP_000564.2
NM_000651.4 517 Missense Mutation CAA,CGA Q126R NP_000642.3

View Full Product Details