Product Details
- SNP ID
-
rs55867570
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:207113036 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGAAGCAGCAGGCCATGCACCCCC[A/C]AAAAACTCCATCTGGGGCTCTTCAT
- Phenotype
-
MIM: 120830
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C4BPA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12072216] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C4BPA
- Gene Name
- complement component 4 binding protein alpha
View Full Product Details