Product Details

SNP ID
rs61785819
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:9103690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAATTGGAAGGAGGGAACCTGT[A/G]TCTGAAGTGATTGACAGGTGGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR157 PubMed Links

Gene Details

Gene
GPR157
Gene Name
G protein-coupled receptor 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024980.4 1634 Intron NP_079256.4
XM_005263496.4 1634 UTR 3 XP_005263553.1
XM_005263497.4 1634 UTR 3 XP_005263554.1

View Full Product Details