Product Details
- SNP ID
-
rs56145411
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:12861478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGCCATCCTGCCTGGCCTGAGCT[A/G]CTGCTCCCAGCTCACCACCTTCTAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF2
PubMed Links
Gene Details
- Gene
- PRAMEF2
- Gene Name
- PRAME family member 2
There are no transcripts associated with this gene.
View Full Product Details