Product Details

SNP ID
rs58748621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:132417677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCAATATGTCCATCTGTACCA[A/G]CCTCTCCAGCCGCTGGACCGTGTTC
Phenotype
MIM: 602886
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR39 PubMed Links

Gene Details

Gene
GPR39
Gene Name
G protein-coupled receptor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001508.2 1104 Missense Mutation AAC,AGC N212S NP_001499.1
XM_011511021.2 1104 Missense Mutation AAC,AGC N212S XP_011509323.1

View Full Product Details