Product Details

SNP ID
rs61754191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27276807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGCAGTGCTGGATCTTAAGAAG[A/G]GCGCCTCACCTGAAGACTTCAAAAA
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 435 Missense Mutation AGC,GGC S27G NP_001290056.1
NM_001303128.1 435 Missense Mutation AGC,GGC S27G NP_001290057.1
NM_173650.2 435 Missense Mutation AGC,GGC S27G NP_775921.1
XM_006711995.3 435 Missense Mutation AGC,GGC S27G XP_006712058.1
XM_017003872.1 435 Missense Mutation AGC,GGC S27G XP_016859361.1
XM_017003873.1 435 Missense Mutation AGC,GGC S27G XP_016859362.1
XM_017003874.1 435 Missense Mutation AGC,GGC S27G XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318949.1 435 Intron NP_001305878.1
NM_001318950.1 435 Intron NP_001305879.1
NM_001318951.1 435 Intron NP_001305880.1
NM_003459.4 435 Intron NP_003450.2
XM_005264547.1 435 Intron XP_005264604.1
XM_005264548.3 435 Intron XP_005264605.1
XM_006712100.2 435 Intron XP_006712163.1
XM_011533102.2 435 Intron XP_011531404.1
XM_011533103.2 435 Intron XP_011531405.1
XM_017004873.1 435 Intron XP_016860362.1
XM_017004874.1 435 Intron XP_016860363.1
XM_017004875.1 435 Intron XP_016860364.1
Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details