Product Details

SNP ID
rs61751580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10141997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCGGCCCGGAGGAACTGGGCGC[C/G]GAGGAGGAGATGGAGGCCGGGCGGC
Phenotype
MIM: 608537
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VHL PubMed Links

Gene Details

Gene
VHL
Gene Name
von Hippel-Lindau tumor suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000551.3 363 Silent Mutation GCC,GCG A50A NP_000542.1
NM_198156.2 363 Silent Mutation GCC,GCG A50A NP_937799.1

View Full Product Details