Product Details

SNP ID
rs57780787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42709695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGGGAGCACCTGGTTGTCAG[C/T]TCTTCCATTTGGGATTCCACGGGCA
Phenotype
MIM: 608116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC13 PubMed Links

Gene Details

Gene
CCDC13
Gene Name
coiled-coil domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144719.3 1459 Silent Mutation GAA,GAG E659E NP_653320.3
XM_005264898.3 1459 Silent Mutation GAA,GAG E618E XP_005264955.1
XM_005264899.4 1459 Intron XP_005264956.1
XM_011533418.2 1459 Silent Mutation GAA,GAG E590E XP_011531720.1
XM_011533419.2 1459 Silent Mutation GAA,GAG E439E XP_011531721.1
XM_011533420.2 1459 Intron XP_011531722.1
XM_011533421.2 1459 Intron XP_011531723.1
XM_017005781.1 1459 Silent Mutation GAA,GAG E439E XP_016861270.1
Gene
HHATL
Gene Name
hedgehog acyltransferase-like
There are no transcripts associated with this gene.

View Full Product Details