Product Details

SNP ID
rs62262473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49011962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACTCAGTGGTGCTATATGACC[C/T]TGTAGTAGGGTGCATCCTGCAAGAG
Phenotype
MIM: 615150 MIM: 612911 MIM: 614584 MIM: 606031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320546.1 383 Missense Mutation CCT,CTT P117L NP_001307475.1
NM_001320547.1 383 Missense Mutation CCT,CTT P92L NP_001307476.1
NM_018031.4 383 Missense Mutation CCT,CTT P173L NP_060501.3

View Full Product Details