Product Details

SNP ID
rs56122560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50278576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACTGGCCAGCATGGGTGCTGT[G/T]GGTGTATCTGCCCACTTGTGAAGCT
Phenotype
MIM: 602725 MIM: 601281
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFRD2 PubMed Links

Gene Details

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
There are no transcripts associated with this gene.

Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304385.1 267 Intron NP_001291314.1
NM_153215.2 267 Intron NP_694947.1
XM_006712979.3 267 UTR 5 XP_006713042.1
XM_006712980.3 267 UTR 5 XP_006713043.1
XM_011533370.2 267 Intron XP_011531672.1
Gene
MIR6872
Gene Name
microRNA 6872
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
There are no transcripts associated with this gene.

View Full Product Details