Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145353.1 | Intron | NP_001138825.1 | |||
NM_172373.3 | Intron | NP_758961.1 | |||
XM_005266276.1 | Intron | XP_005266333.1 | |||
XM_005266277.3 | Intron | XP_005266334.1 | |||
XM_011534950.1 | Intron | XP_011533252.1 | |||
XM_011534951.2 | Intron | XP_011533253.1 | |||
XM_017020409.1 | Intron | XP_016875898.1 | |||
XM_017020410.1 | Intron | XP_016875899.1 | |||
XM_017020411.1 | Intron | XP_016875900.1 | |||
XM_017020412.1 | Intron | XP_016875901.1 | |||
XM_017020413.1 | Intron | XP_016875902.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_007187.3 | Intron | NP_009118.1 | |||
XM_005266245.2 | Intron | XP_005266302.1 | |||
XM_017020380.1 | Intron | XP_016875869.1 |