Product Details

SNP ID
rs57510477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46127203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTGTGATGTTTCCCCTCCTGG[C/T]CCTCATCTTGAACAAAGAAAAGGGA
Phenotype
MIM: 153430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCP1 PubMed Links

Gene Details

Gene
LCP1
Gene Name
lymphocyte cytosolic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002298.4 2408 UTR 3 NP_002289.2
XM_005266374.1 2408 UTR 3 XP_005266431.1

View Full Product Details