Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001111298.2 | 3974 | UTR 3 | NP_001104768.2 | ||
NM_001286360.1 | 3974 | UTR 3 | NP_001273289.1 | ||
NM_001286361.1 | 3974 | UTR 3 | NP_001273290.1 | ||
NM_173672.4 | 3974 | UTR 3 | NP_775943.1 | ||
XM_011535765.2 | 3974 | Intron | XP_011534067.1 | ||
XM_011535766.2 | 3974 | Intron | XP_011534068.1 | ||
XM_011535767.2 | 3974 | Intron | XP_011534069.1 | ||
XM_011535769.2 | 3974 | Intron | XP_011534071.1 | ||
XM_017010774.1 | 3974 | Intron | XP_016866263.1 |