Product Details

SNP ID
rs61991696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60749793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTTGTTCCTGAGACAGCAAATA[A/G]AATGAGAATAATTGAGTGAAAATTA
Phenotype
MIM: 602659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MNAT1 PubMed Links

Gene Details

Gene
MNAT1
Gene Name
MNAT1, CDK activating kinase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177963.1 Intron NP_001171434.1
NM_002431.3 Intron NP_002422.1
XM_005267687.2 Intron XP_005267744.1
XM_005267688.2 Intron XP_005267745.1
XM_017021332.1 Intron XP_016876821.1
XM_017021333.1 Intron XP_016876822.1
XM_017021334.1 Intron XP_016876823.1

View Full Product Details