Product Details

SNP ID
rs60979440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64923513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTACTTTTGTTTATTTTGTATT[A/G]TGATCAAGGATCAAAACTATTTGAA
Phenotype
MIM: 608577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHURC1 PubMed Links

Gene Details

Gene
CHURC1
Gene Name
churchill domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204063.1 Intron NP_001190992.1
NM_001204064.1 Intron NP_001190993.1
NM_145165.3 Intron NP_660148.3
Gene
CHURC1-FNTB
Gene Name
CHURC1-FNTB readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202558.1 Intron NP_001189487.1
NM_001202559.1 Intron NP_001189488.1

View Full Product Details