Product Details

SNP ID
rs55651033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24369731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGCTGGGGGTGGCCGTGTTCT[C/T]GAGTGTCCCAGCATCCGCATCACCT
Phenotype
MIM: 602699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFATC4 PubMed Links

Gene Details

Gene
NFATC4
Gene Name
nuclear factor of activated T-cells 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136022.2 654 Silent Mutation CTC,CTT L174L NP_001129494.1
NM_001198965.1 654 Silent Mutation CTC,CTT L111L NP_001185894.1
NM_001198966.2 654 Silent Mutation CTC,CTT L41L NP_001185895.1
NM_001198967.2 654 Silent Mutation CTC,CTT L174L NP_001185896.1
NM_001288802.1 654 Silent Mutation CTC,CTT L99L NP_001275731.1
NM_001320043.1 654 Silent Mutation CTC,CTT L174L NP_001306972.1
NM_004554.4 654 Silent Mutation CTC,CTT L111L NP_004545.2
XM_011536797.2 654 Silent Mutation CTC,CTT L99L XP_011535099.1
XM_011536798.1 654 Silent Mutation CTC,CTT L88L XP_011535100.1
XM_011536799.2 654 Silent Mutation CTC,CTT L99L XP_011535101.1

View Full Product Details