Product Details

SNP ID
rs55819223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94285634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTTGTGGATTGGGCAGCTCTT[C/T]AAAGAAATGGTGAGTTTGATGTCAC
Phenotype
MIM: 616790 MIM: 605271
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP4R4 PubMed Links

Gene Details

Gene
PPP4R4
Gene Name
protein phosphatase 4 regulatory subunit 4
There are no transcripts associated with this gene.

Gene
SERPINA10
Gene Name
serpin family A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100607.2 Intron NP_001094077.1
NM_016186.2 Intron NP_057270.1
XM_005267733.4 Intron XP_005267790.1
XM_017021353.1 Intron XP_016876842.1

View Full Product Details