Product Details

SNP ID
rs60890987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97430733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGCCTAATCTCACTGAACTTA[C/T]AATTCATGATAAAATAGTTAATTGT
Phenotype
MIM: 606493 MIM: 172250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC1 PubMed Links

Gene Details

Gene
EXOSC1
Gene Name
exosome component 1
There are no transcripts associated with this gene.

Gene
LOC644215
Gene Name
uncharacterized LOC644215
There are no transcripts associated with this gene.

Gene
PGAM1
Gene Name
phosphoglycerate mutase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317079.1 Intron NP_001304008.1
NM_002629.3 Intron NP_002620.1

View Full Product Details