Product Details

SNP ID
rs56323538
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:45345125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTGGTGGGGGCCCATCAGATAG[C/T]GATTCTTCCCGGCCCACAAATGGAC
Phenotype
MIM: 604933 MIM: 604746 MIM: 613931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUTYH PubMed Links

Gene Details

Gene
MUTYH
Gene Name
mutY DNA glycosylase
There are no transcripts associated with this gene.

Gene
TESK2
Gene Name
testis-specific kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320800.1 2162 Silent Mutation TCA,TCG S394S NP_001307729.1
NM_007170.2 2162 Silent Mutation TCA,TCG S477S NP_009101.2
XM_005270355.3 2162 Silent Mutation TCA,TCG S428S XP_005270412.1
XM_006710287.2 2162 Silent Mutation TCA,TCG S477S XP_006710350.1
XM_011540497.1 2162 Silent Mutation TCA,TCG S477S XP_011538799.1
XM_011540500.2 2162 Intron XP_011538802.1
XM_017000047.1 2162 Silent Mutation TCA,TCG S428S XP_016855536.1
XM_017000048.1 2162 Silent Mutation TCA,TCG S269S XP_016855537.1
XM_017000049.1 2162 Silent Mutation TCA,TCG S269S XP_016855538.1
Gene
TOE1
Gene Name
target of EGR1, member 1 (nuclear)
There are no transcripts associated with this gene.

View Full Product Details