Product Details
- SNP ID
-
rs56004824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:66798196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGAAGGTTTTGCGCTGGATTTTC[C/T]TCAGAAAACTCACGTTTATGTGGGA
- Phenotype
-
MIM: 606413
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
INSL5
PubMed Links
Gene Details
- Gene
- INSL5
- Gene Name
- insulin like 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005478.4 |
263 |
Silent Mutation |
GAA,GAG |
E75E |
NP_005469.2 |
View Full Product Details