Product Details

SNP ID
rs61776918
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:35724793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCAGATTGGAACGAACATTATT[C/T]CCCAGAATTGGTTCATCTTGGGAAA
Phenotype
MIM: 605434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf216 PubMed Links

Gene Details

Gene
C1orf216
Gene Name
chromosome 1 open reading frame 216
There are no transcripts associated with this gene.

Gene
CLSPN
Gene Name
claspin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190481.1 Intron NP_001177410.1
NM_022111.3 Intron NP_071394.2
XM_011541938.2 Intron XP_011540240.1
XM_011541939.2 Intron XP_011540241.1
XM_011541940.2 Intron XP_011540242.1
XM_011541941.2 Intron XP_011540243.1

View Full Product Details