Product Details

SNP ID
rs60236485
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65499935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGCTAAGACAAGTATTGGAGAA[A/G]TATAGAAGATAGAAAAATATAAAGC
Phenotype
MIM: 607924
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MALAT1 PubMed Links
Additional Information
For this assay, SNP(s) [rs148394780] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MALAT1
Gene Name
metastasis associated lung adenocarcinoma transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details