Product Details

SNP ID
rs61896975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55639048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGAGCATACTCTGCAGAGAGA[C/T]GCAGCAAAGCTCTTGCCACTTGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 691 Missense Mutation CGC,TGC R231C NP_001004124.1

View Full Product Details