Product Details

SNP ID
hCV90501224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62765183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGATCACTCATTTTTTTTCTAGG[C/T]TGGACTCTTCACAGAAATTGGGCCC
Phenotype
MIM: 602013 MIM: 602946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR2G PubMed Links

Gene Details

Gene
POLR2G
Gene Name
RNA polymerase II subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002696.2 394 Missense Mutation GCT,GTT A95V NP_002687.1
Gene
TAF6L
Gene Name
TATA-box binding protein associated factor 6 like
There are no transcripts associated with this gene.

View Full Product Details