Product Details

SNP ID
rs55983299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85961083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTTAAGAATTTTAATGTGTGTA[G/T]GGGGGGTGGTGGGGGGATAGGGGAG
Phenotype
MIM: 603025
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PICALM PubMed Links

Gene Details

Gene
PICALM
Gene Name
phosphatidylinositol binding clathrin assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008660.2 Intron NP_001008660.1
NM_001206946.1 Intron NP_001193875.1
NM_001206947.1 Intron NP_001193876.1
NM_007166.3 Intron NP_009097.2
XM_005274322.2 Intron XP_005274379.1
XM_005274323.2 Intron XP_005274380.1
XM_005274324.2 Intron XP_005274381.1
XM_005274325.2 Intron XP_005274382.1
XM_005274326.2 Intron XP_005274383.1
XM_005274327.2 Intron XP_005274384.1
XM_005274328.2 Intron XP_005274385.1
XM_005274329.3 Intron XP_005274386.1
XM_005274330.2 Intron XP_005274387.1
XM_005274331.2 Intron XP_005274388.1
XM_005274332.2 Intron XP_005274389.1
XM_005274333.2 Intron XP_005274390.1
XM_005274334.2 Intron XP_005274391.1
XM_005274335.2 Intron XP_005274392.1
XM_005274336.2 Intron XP_005274393.1
XM_005274337.2 Intron XP_005274394.1
XM_005274340.2 Intron XP_005274397.1
XM_006718699.2 Intron XP_006718762.1
XM_006718700.2 Intron XP_006718763.1
XM_006718701.3 Intron XP_006718764.1
XM_011545293.1 Intron XP_011543595.1
XM_017018381.1 Intron XP_016873870.1
XM_017018382.1 Intron XP_016873871.1
XM_017018383.1 Intron XP_016873872.1
XM_017018384.1 Intron XP_016873873.1
XM_017018385.1 Intron XP_016873874.1
XM_017018386.1 Intron XP_016873875.1
XM_017018387.1 Intron XP_016873876.1
XM_017018388.1 Intron XP_016873877.1

View Full Product Details