Product Details

SNP ID
hCV90575389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124801896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACAGCGCCAACAGCAACGCG[G/T]ACCACGTAGGTGCCGGGCCCCCTGC
Phenotype
MIM: 611144
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM175B PubMed Links

Gene Details

Gene
FAM175B
Gene Name
family with sequence similarity 175 member B
There are no transcripts associated with this gene.

Gene
METTL10
Gene Name
methyltransferase like 10
There are no transcripts associated with this gene.

View Full Product Details