Product Details

SNP ID
rs61876180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:204100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGTGGGGGGACAGTGATCCCA[C/T]AGTCCCCAGGGAGTGACTGGGAGGC
Phenotype
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 610176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BET1L PubMed Links

Gene Details

Gene
BET1L
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098787.1 1780 UTR 3 NP_001092257.1
NM_016526.4 1780 UTR 3 NP_057610.2
Gene
MIR6743
Gene Name
microRNA 6743
There are no transcripts associated with this gene.

Gene
ODF3
Gene Name
outer dense fiber of sperm tails 3
There are no transcripts associated with this gene.

Gene
RIC8A
Gene Name
RIC8 guanine nucleotide exchange factor A
There are no transcripts associated with this gene.

Gene
SCGB1C1
Gene Name
secretoglobin family 1C member 1
There are no transcripts associated with this gene.

View Full Product Details