Product Details
- SNP ID
-
rs72704611
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:150745012 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTTTCATTTTAAAAAGCTATCAC[C/T]GGTCTACACCCAAGCTGATCAGTGC
- Phenotype
-
MIM: 116845
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTSS
PubMed Links
Gene Details
- Gene
- CTSS
- Gene Name
- cathepsin S
View Full Product Details