Product Details

SNP ID
rs72702899
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:152786439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGACAGAAAGAATAACTGGTT[G/T]GCTGTGCCTTTGGGCTCCAAAAAAA
Phenotype
MIM: 612607 MIM: 612608
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LCE1E PubMed Links

Gene Details

Gene
LCE1E
Gene Name
late cornified envelope 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178353.1 Intron NP_848130.1
Gene
LCE1F
Gene Name
late cornified envelope 1F
There are no transcripts associated with this gene.

View Full Product Details