Product Details

SNP ID
rs73010454
Assay Type
Functionally Tested
NCBI dbSNP Submissions
23
Location
Chr.1:152971913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATGTCTTCCCAGCAGCAGCAGC[A/G]GCAGCAGCAGCAGTGCCCACCCCAG
Phenotype
MIM: 616363
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 781 Missense Mutation CAG,CGG Q8R NP_775103.1
XM_017000482.1 781 Missense Mutation CAG,CGG Q8R XP_016855971.1

View Full Product Details