Product Details

SNP ID
rs73008417
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:153204362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTCTAAATGAATTAAATACAA[C/T]ATATGGAGTTACAGAGACACCAACT
Phenotype
MIM: 611042
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LELP1 PubMed Links

Gene Details

Gene
LELP1
Gene Name
late cornified envelope like proline rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010857.2 Intron NP_001010857.1
Gene
SPRR2G
Gene Name
small proline rich protein 2G
There are no transcripts associated with this gene.

View Full Product Details