Product Details

SNP ID
rs74153217
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:247965871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACATACATTGTATGTGTTTCA[A/G]CTTCCATTCTGTAGGTCTCGGCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2AK2 PubMed Links

Gene Details

Gene
OR2AK2
Gene Name
olfactory receptor family 2 subfamily AK member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004491.1 540 Silent Mutation CAA,CAG Q180Q NP_001004491.1
Gene
OR2L13
Gene Name
olfactory receptor family 2 subfamily L member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304535.1 540 Intron NP_001291464.1
NM_175911.3 540 Intron NP_787107.1
XM_011544169.2 540 Intron XP_011542471.1

View Full Product Details