Product Details

SNP ID
rs73117165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:242000080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAAAAGCTGACCCTGAAAAAT[C/T]ATCTTATGGTCACATTTAGTTTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01237 PubMed Links

Gene Details

Gene
LINC01237
Gene Name
long intergenic non-protein coding RNA 1237
There are no transcripts associated with this gene.

Gene
LOC285097
Gene Name
uncharacterized FLJ38379
There are no transcripts associated with this gene.

View Full Product Details