Product Details

SNP ID
rs72961101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127197363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCTGGACCCGGGCTGCTCCCG[A/C]TCCTCCTCCCGGTCACCCTGCAGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C3orf56 PubMed Links

Gene Details

Gene
C3orf56
Gene Name
chromosome 3 open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007534.2 918 Silent Mutation CGA,CGC R226R NP_001007535.1

View Full Product Details