Product Details
- SNP ID
-
rs66491615
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:6312124 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGAACAATGCATCTTGCTACCA[C/G]AGCACATCGGAAGCATCCCTTCAGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ33360
PubMed Links
Gene Details
- Gene
- FLJ33360
- Gene Name
- FLJ33360 protein
There are no transcripts associated with this gene.
View Full Product Details