Product Details

SNP ID
rs73122560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75028178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGAGTCTTTCCTCCTCTGCTCT[A/G]ATGGTTAAGAAATGTCTTGAAAGCT
Phenotype
MIM: 616782
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCNT4 PubMed Links

Gene Details

Gene
GCNT4
Gene Name
glucosaminyl (N-acetyl) transferase 4, core 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016591.2 2722 UTR 3 NP_057675.1

View Full Product Details