Product Details

SNP ID
rs72912443
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:86943612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAAACTGCACCTTGTGTTATAC[C/T]ACTAGGGGGTAACACAGCATGGGGC
Phenotype
MIM: 182132
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HTR1E PubMed Links

Gene Details

Gene
HTR1E
Gene Name
5-hydroxytryptamine receptor 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000865.2 Intron NP_000856.1
XM_011535789.2 Intron XP_011534091.1

View Full Product Details