Product Details

SNP ID
rs67790793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49684485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGTAGAACTCACAGATATGGAG[A/G]ACCGACTATATAAGGAATTCAGCAT
Phenotype
MIM: 187430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRISP2 PubMed Links

Gene Details

Gene
CRISP2
Gene Name
cysteine rich secretory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142407.2 1023 Intron NP_001135879.1
NM_001142408.2 1023 Intron NP_001135880.1
NM_001142417.2 1023 Intron NP_001135889.1
NM_001142435.2 1023 Intron NP_001135907.1
NM_001261822.1 1023 Intron NP_001248751.1
NM_003296.3 1023 Intron NP_003287.1
XM_005249349.1 1023 Intron XP_005249406.1
XM_005249350.1 1023 Intron XP_005249407.1
XM_005249351.3 1023 Intron XP_005249408.1
XM_005249352.3 1023 Intron XP_005249409.1
XM_005249353.3 1023 Intron XP_005249410.1
XM_005249356.1 1023 Intron XP_005249413.1
XM_011514841.1 1023 Intron XP_011513143.1
XM_011514842.1 1023 Intron XP_011513144.1
XM_011514843.2 1023 UTR 3 XP_011513145.1
XM_011514844.1 1023 Intron XP_011513146.1

View Full Product Details