Product Details

SNP ID
rs73088329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:24797901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATAAAATGTCCTAGAGACATA[A/C]ATTTGGGGGAAAGATGCTTTTGAGA
Phenotype
MIM: 606732
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OSBPL3 PubMed Links

Gene Details

Gene
OSBPL3
Gene Name
oxysterol binding protein like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015550.3 5320 UTR 3 NP_056365.1
NM_145320.2 5320 UTR 3 NP_663160.1
NM_145321.2 5320 UTR 3 NP_663161.1
NM_145322.2 5320 UTR 3 NP_663162.1
XM_005249698.3 5320 UTR 3 XP_005249755.1
XM_006715681.3 5320 UTR 3 XP_006715744.1
XM_006715682.3 5320 UTR 3 XP_006715745.1
XM_006715683.3 5320 UTR 3 XP_006715746.1
XM_006715684.3 5320 UTR 3 XP_006715747.1
XM_011515258.2 5320 UTR 3 XP_011513560.1
XM_011515259.2 5320 UTR 3 XP_011513561.1
XM_017011948.1 5320 UTR 3 XP_016867437.1
XM_017011949.1 5320 UTR 3 XP_016867438.1

View Full Product Details