Product Details

SNP ID
rs71528572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29566657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGTGCGCGCCACGCTGCTCCCG[A/G]AGGCGGGCAGGTCCCCGGAGGAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRR15 PubMed Links

Gene Details

Gene
PRR15
Gene Name
proline rich 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175887.2 1843 Missense Mutation AAG,GAG K110E NP_787083.1
XM_005249653.3 1843 Missense Mutation AAG,GAG K110E XP_005249710.1
XM_011515198.1 1843 Missense Mutation AAG,GAG K110E XP_011513500.1
XM_011515199.1 1843 Missense Mutation AAG,GAG K110E XP_011513501.1
XM_017011842.1 1843 Missense Mutation AAG,GAG K110E XP_016867331.1

View Full Product Details