Product Details

SNP ID
rs73102277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36396612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTAGAACCTATAGGGTTGGAA[C/T]GGTTTCTTCTAAAAAATATTTTAAA
Phenotype
MIM: 616027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANLN PubMed Links

Gene Details

Gene
ANLN
Gene Name
anillin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284301.2 Intron NP_001271230.1
NM_001284302.2 Intron NP_001271231.1
NM_018685.4 Intron NP_061155.2
XM_006715746.1 Intron XP_006715809.1
XM_006715747.3 Intron XP_006715810.1
XM_017012354.1 Intron XP_016867843.1
XM_017012355.1 Intron XP_016867844.1
XM_017012356.1 Intron XP_016867845.1
Gene
KIAA0895
Gene Name
KIAA0895
There are no transcripts associated with this gene.

View Full Product Details