Product Details
- SNP ID
-
rs73463627
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149842900 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGAGTGGAGCTTGTCTTCCAAT[C/T]GGTTTTTGGAAATATTTCATTTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSPO
PubMed Links
Gene Details
- Gene
- SSPO
- Gene Name
- SCO-spondin
There are no transcripts associated with this gene.
- Gene
- ZNF862
- Gene Name
- zinc finger protein 862
View Full Product Details