Product Details

SNP ID
rs73263786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69806576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCCTGGTCTGGGACCCTTAA[C/T]GAGCACTCCAGCTGCCTGTTATTTG
Phenotype
MIM: 120350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL13A1 PubMed Links

Gene Details

Gene
COL13A1
Gene Name
collagen type XIII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130103.1 Intron NP_001123575.1
NM_001320951.1 Intron NP_001307880.1
NM_080798.3 Intron NP_542988.3
NM_080800.3 Intron NP_542990.3
NM_080801.3 Intron NP_542991.3
NM_080802.3 Intron NP_542992.3
NM_080805.3 Intron NP_542995.3
XM_011539292.2 Intron XP_011537594.1
XM_011539293.2 Intron XP_011537595.1
XM_011539294.2 Intron XP_011537596.1
XM_011539295.2 Intron XP_011537597.1
XM_017015676.1 Intron XP_016871165.1
XM_017015677.1 Intron XP_016871166.1
XM_017015678.1 Intron XP_016871167.1
XM_017015679.1 Intron XP_016871168.1
XM_017015680.1 Intron XP_016871169.1
XM_017015681.1 Intron XP_016871170.1
XM_017015682.1 Intron XP_016871171.1
XM_017015683.1 Intron XP_016871172.1
XM_017015684.1 Intron XP_016871173.1
XM_017015685.1 Intron XP_016871174.1
XM_017015686.1 Intron XP_016871175.1
XM_017015687.1 Intron XP_016871176.1
XM_017015688.1 Intron XP_016871177.1
XM_017015689.1 Intron XP_016871178.1
XM_017015690.1 Intron XP_016871179.1
XM_017015691.1 Intron XP_016871180.1
XM_017015692.1 Intron XP_016871181.1
XM_017015693.1 Intron XP_016871182.1
XM_017015694.1 Intron XP_016871183.1
XM_017015695.1 Intron XP_016871184.1
XM_017015696.1 Intron XP_016871185.1
XM_017015697.1 Intron XP_016871186.1

View Full Product Details