Product Details

SNP ID
rs73434269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18173705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTGTCCTGCTCTGGGGCCTGT[C/T]CCTGCTGTTTAGTATGCTGGAGTGG
Phenotype
MIM: 607230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRGPRX4 PubMed Links

Gene Details

Gene
MRGPRX4
Gene Name
MAS related GPR family member X4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054032.3 Intron NP_473373.2

View Full Product Details