Product Details

SNP ID
rs71448606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21528388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTGTTCTTTTTCATGATAAAC[C/T]TTTCCATCGTATTGTATTAACAAAT
Phenotype
MIM: 615078 MIM: 138571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLT1B PubMed Links

Gene Details

Gene
GOLT1B
Gene Name
golgi transport 1B
There are no transcripts associated with this gene.

Gene
GYS2
Gene Name
glycogen synthase 2
There are no transcripts associated with this gene.

Gene
SPX
Gene Name
spexin hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030572.3 Intron NP_085049.1

View Full Product Details