Product Details
- SNP ID
-
rs71647806
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:89776730 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGAGCCTGACCAAGATCGAGACG[C/G]TGCGCCTGGCCATCCGCTACATCGG
- Phenotype
-
MIM: 151530
MIM: 605195
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANPEP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28462216] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANPEP
- Gene Name
- alanyl aminopeptidase, membrane
There are no transcripts associated with this gene.
- Gene
- MESP2
- Gene Name
- mesoderm posterior bHLH transcription factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039958.1 |
373 |
Missense Mutation |
CTG,GTG |
L125V |
NP_001035047.1 |
View Full Product Details